Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023229.2 | NP_075718.2 | 3 - 4 | 763 | 73 | |
GenBank mRNA | AF168682.1 | - | 2 - 3 | 658 | 73 | |
AK002552.1 | - | 3 - 4 | 714 | 73 | ||
AK038596.1 | - | 3 - 4 | 715 | 73 | ||
AK052667.1 | - | 3 - 4 | 708 | 73 | ||
AK139374.1 | - | 2 - 3 | 1481 | 73 | ||
AK151859.1 | - | 3 - 4 | 715 | 73 | ||
AK152369.1 | - | 3 - 4 | 764 | 73 | ||
AK159751.1 | - | 3 - 4 | 715 | 73 | ||
AK164317.1 | - | 721 | 73 | |||
AK167346.1 | - | 2 - 3 | 1475 | 73 | ||
BC013547.1 | - | 3 - 4 | 680 | 4218374 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |