Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700017877.mm.1 |
TC0700002033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700017877.mm.1 |
NM_145387.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700017877.mm.1 |
NM_145387.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700017880.mm.1 |
TC0700002033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700017880.mm.1 |
NM_145387.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700017880.mm.1 |
NM_145387.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700017878.mm.1 |
TC0700002033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700017878.mm.1 |
NM_145387.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700017878.mm.1 |
NM_145387.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700017879.mm.1 |
TC0700002033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700017879.mm.1 |
NM_145387.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700017879.mm.1 |
NM_145387.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700017881.mm.1 |
TC0700002033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700017881.mm.1 |
NM_145387.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700017881.mm.1 |
NM_145387.4 |
Refseq |