Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nobox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130869.3 | NP_570939.1 | 2 - 3 | 357 | 72 | |
GenBank mRNA | AB221582.1 | - | 2 - 3 | 334 | 72 | |
AY061761.1 | - | 2 - 3 | 365 | 72 | ||
BC125280.1 | - | 2 - 3 | 353 | 40129795 | 72 | |
BC125282.1 | - | 2 - 3 | 353 | 40129797 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |