Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289656.1 | NP_001276585.1 | 11 - 12 | 2423 | 99 | |
NM_001289657.1 | NP_001276586.1 | 10 - 11 | 2397 | 99 | ||
NM_178630.4 | NP_848745.2 | 10 - 11 | 2408 | 99 | ||
GenBank mRNA | AK029793.1 | - | 11 - 12 | 2424 | 99 | |
AK029798.1 | - | 10 - 11 | 1981 | 99 | ||
DQ867031.1 | - | 10 - 11 | 2409 | 99 | ||
DQ867032.1 | - | 10 - 11 | 2409 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |