Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030727.5 | NP_109652.3 | 5 - 6 | 635 | 100 | |
GenBank mRNA | AF529192.1 | - | 5 - 6 | 635 | 100 | |
AY024359.2 | - | 3 - 4 | 401 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mus musculus Probe spans exons |