Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153537.4 | NP_705765.3 | 6 - 7 | 1933 | 83 | |
GenBank mRNA | AK122336.1 | - | 3 - 4 | 1651 | 83 | |
BC058712.1 | - | 6 - 7 | 1929 | 6842596 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |