Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supv3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181423.2 | NP_852088.1 | 8 - 9 | 1027 | 87 | |
GenBank mRNA | AJ968954.1 | - | 8 - 9 | 1054 | 87 | |
AK031911.1 | - | 8 - 9 | 1034 | 87 | ||
AK143755.1 | - | 8 - 9 | 1052 | 87 | ||
BC049796.1 | - | 8 - 9 | 1027 | 6332552 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |