Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177372.3 | NP_796346.2 | 12 - 13 | 1942 | 67 | |
GenBank mRNA | AK028381.1 | - | 12 - 13 | 1919 | 67 | |
BC030382.1 | - | 12 - 13 | 1883 | 67 | ||
BC115716.1 | - | 12 - 13 | 1896 | 40085888 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |