Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081127.1 | NP_001074596.1 | 20 - 21 | 3227 | 70 | |
GenBank mRNA | AK137573.1 | - | 5 - 6 | 679 | 70 | |
BC158014.1 | - | 20 - 21 | 3060 | 9088151 | 70 | |
BC172106.1 | - | 20 - 21 | 3043 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |