Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bud13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146000.2 | NP_666112.1 | 1 - 2 | 159 | 80 | |
GenBank mRNA | AK037134.1 | - | 1 - 2 | 156 | 80 | |
AK038126.1 | - | 1 - 2 | 164 | 80 | ||
AK051047.1 | - | 1 - 2 | 164 | 80 | ||
AK077744.1 | - | 1 - 2 | 164 | 80 | ||
BC025490.1 | - | 1 - 2 | 160 | 5252666 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |