Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177326.3 | NP_796300.1 | 10 - 11 | 1215 | 139 | |
GenBank mRNA | AK037275.1 | - | 10 - 11 | 1217 | 139 | |
AY167030.1 | - | 9 - 10 | 941 | 139 | ||
BC063068.1 | - | 1203 | 139 | |||
BC086650.1 | - | 10 - 11 | 1146 | 6849008 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |