Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26471089 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26471088 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26471086 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2124891 |
EF102773.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2124891 |
NM_001093749.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P383530 |
AK155483.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P383530 |
AK041037.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P383530 |
AK049878.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P383530 |
NM_176993.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P383530 |
AK155483.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P383530 |
AK041037.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P383530 |
AK049878.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P383530 |
NM_176993.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2076550 |
EF102773.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2076550 |
NM_001093749.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
71457 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
536919 |
AK141534.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
536919 |
AK170522.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
536919 |
BC023471.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
536919 |
NM_177084.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
675295 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932818 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
603136 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
72712 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
739678 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301060 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
948117 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
581380 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
878245 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
285011 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
88608 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
162901 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P174121 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P383530 |
AK155483.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P383530 |
AK041037.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P383530 |
AK049878.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P383530 |
NM_176993.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P383530 |
AK155483.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P383530 |
AK041037.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P383530 |
AK049878.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P383530 |
NM_176993.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2076550 |
EF102773.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2076550 |
NM_001093749.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2124891 |
EF102773.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2124891 |
NM_001093749.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471103 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471082 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471095 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471081 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471102 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471078 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471085 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471090 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471091 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471079 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471088 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471083 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471100 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471097 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471099 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471092 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26471093 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06771174 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P174121 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P383530 |
AK155483.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P383530 |
AK041037.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P383530 |
AK049878.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P383530 |
NM_176993.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P383530 |
AK155483.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P383530 |
AK041037.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P383530 |
AK049878.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P383530 |
NM_176993.3 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115049 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P174121 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07621551 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07621559 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07621548 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07621546 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07621553 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26471086 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26471088 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26471089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471157 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471185 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471204 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471191 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471263 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471236 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471268 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471081 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471245 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471219 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471167 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471251 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471223 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471261 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471267 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471166 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471165 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471200 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471196 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471135 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471227 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471271 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471107 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471163 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26471232 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471088 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471197 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471100 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471082 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471112 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07679979 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471131 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471090 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471133 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471106 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471190 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07993836 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08135838 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471137 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471083 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471081 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471135 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471105 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471095 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471085 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07528619 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471079 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471092 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471093 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07857830 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471099 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471110 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07993840 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471091 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07749255 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471125 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471097 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471078 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07993844 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471193 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471116 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471103 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471138 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471118 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08013368 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471108 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26471120 |
- |
- |