Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sesn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030261.4 | NP_084537.2 | 6 - 7 | 1184 | 70 | |
GenBank mRNA | AK017464.1 | - | 6 - 7 | 1081 | 70 | |
AK155398.1 | - | 6 - 7 | 1185 | 70 | ||
BC132086.1 | - | 6 - 7 | 1005 | 40130363 | 70 | |
BC138690.1 | - | 6 - 7 | 1005 | 8861712 | 70 | |
KF543243.1 | - | 6 - 7 | 933 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |