Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018758.2 | NP_061228.1 | 1 - 2 | 145 | 72 | |
GenBank mRNA | AF070975.1 | - | 1 - 2 | 99 | 72 | |
AK045965.1 | - | 1 - 2 | 146 | 72 | ||
AK082046.1 | - | 65 | 72 | |||
AK169462.1 | - | 1 - 2 | 131 | 72 | ||
AK170182.1 | - | 1 - 2 | 93 | 72 | ||
BC005423.1 | - | 1 - 2 | 77 | 3487448 | 72 | |
BC009666.1 | - | 1 - 2 | 89 | 3490710 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |