Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009200.3 | NP_033226.1 | 7 - 8 | 1353 | 62 | |
GenBank mRNA | AK159197.1 | - | 7 - 8 | 1354 | 62 | |
BC138120.1 | - | 7 - 8 | 1255 | 8861139 | 62 | |
BC138140.1 | - | 7 - 8 | 1255 | 8861159 | 62 | |
D83262.1 | - | 7 - 8 | 1347 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |