Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164376.1 | NP_001157848.1 | 9 - 10 | 1436 | 105 | |
NM_001164519.1 | NP_001157991.1 | 4 - 5 | 595 | 105 | ||
NM_177612.3 | NP_808280.2 | 9 - 10 | 1364 | 105 | ||
GenBank mRNA | AF344871.1 | - | 9 - 10 | 1436 | 105 | |
AK142745.1 | - | 9 - 10 | 1437 | 105 | ||
BC112409.1 | - | 9 - 10 | 1338 | 40060582 | 105 | |
DQ380429.1 | - | 9 - 10 | 1364 | 105 | ||
DQ380430.1 | - | 4 - 5 | 594 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |