Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172772.2 | NP_766360.2 | 2 - 3 | 960 | 276 | |
GenBank mRNA | AK035930.1 | - | 2 - 3 | 961 | 276 | |
AK046403.1 | - | 2 - 3 | 947 | 276 | ||
AK046623.1 | - | 2 - 3 | 934 | 276 | ||
AK220158.1 | - | 2 - 3 | 989 | 276 | ||
BC049357.1 | - | 2 - 3 | 937 | 276 | ||
BC055816.1 | - | 2 - 3 | 688 | 276 | ||
BC058683.1 | - | 2 - 3 | 918 | 6834871 | 276 | |
BC069845.1 | - | 2 - 3 | 673 | 276 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |