Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB186125.1 | - | 5 - 6 | 2441 | 70 | |
AK173252.1 | - | 4 - 5 | 982 | 70 | ||
BC139006.1 | - | 4 - 5 | 1913 | 8862028 | 70 | |
BC158061.1 | - | 4 - 5 | 1916 | 9007361 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |