Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122873.1 | NP_001116345.1 | 5 - 6 | 577 | 69 | |
NM_027086.3 | NP_081362.2 | 5 - 6 | 525 | 69 | ||
GenBank mRNA | AK009025.1 | - | 5 - 6 | 526 | 69 | |
AK154496.1 | - | 5 - 6 | 535 | 69 | ||
AK179100.1 | - | 1 - 2 | 89 | 69 | ||
AK182178.1 | - | 1 - 2 | 89 | 69 | ||
AK183337.1 | - | 1 - 2 | 69 | |||
AK207226.1 | - | 1 - 2 | 89 | 69 | ||
BC016456.1 | - | 4 - 5 | 511 | 4912715 | 69 | |
BC094047.1 | - | 4 - 5 | 514 | 5253101 | 69 | |
CX203425.1 | - | 5 - 6 | 555 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |