Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033498.1 | NP_001028670.1 | 8 - 9 | 951 | 83 | |
GenBank mRNA | AK040545.1 | - | 9 - 10 | 840 | 83 | |
AK041631.1 | - | 8 - 9 | 951 | 83 | ||
BC138948.1 | - | 8 - 9 | 768 | 8861970 | 83 | |
BC138949.1 | - | 8 - 9 | 768 | 8861971 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |