Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igdcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008988.2 | NP_033014.1 | 9 - 10 | 1821 | 83 | |
GenBank mRNA | AF026465.1 | - | 9 - 10 | 1821 | 83 | |
AK011916.1 | - | 9 - 10 | 1859 | 83 | ||
AK051027.1 | - | 9 - 10 | 1822 | 83 | ||
BC053057.1 | - | 9 - 10 | 1798 | 5698028 | 83 | |
BC086481.1 | - | 9 - 10 | 1805 | 6505546 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |