Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdx1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039824.2 | NP_001034913.1 | 1 - 2 | 130 | 107 | |
GenBank mRNA | AK008401.2 | - | 1 - 2 | 128 | 107 | |
AK010211.1 | - | 1 - 2 | 101 | 107 | ||
AK020979.1 | - | 1 - 2 | 134 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |