Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024139.1 | NP_001019310.1 | 3 - 4 | 1474 | 79 | |
GenBank mRNA | BC043308.1 | - | 1 - 2 | 95 | 79 | |
BC094677.1 | - | 3 - 4 | 1474 | 30619053 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |