Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028874.2 | NP_083150.1 | 7 - 8 | 2698 | 58 | |
GenBank mRNA | AK014388.1 | - | 1 - 2 | 145 | 58 | |
AK032107.1 | - | 3 - 4 | 336 | 58 | ||
AK044307.1 | - | 6 - 7 | 1742 | 58 | ||
AK122231.1 | - | 7 - 8 | 2919 | 58 | ||
BC004635.1 | - | 7 - 8 | 1170 | 58 | ||
BC051164.1 | - | 7 - 8 | 2918 | 58 | ||
BC063262.1 | - | 7 - 8 | 2698 | 6405897 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |