Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011176.4 | NP_035306.2 | 8 - 9 | 1188 | 73 | |
GenBank mRNA | AF042822.3 | - | 8 - 9 | 1077 | 73 | |
AK085410.1 | - | 6 - 7 | 1614 | 73 | ||
AK166745.1 | - | 8 - 9 | 1179 | 73 | ||
BC005496.1 | - | 8 - 9 | 1187 | 3488059 | 73 | |
BC008514.1 | - | 8 - 9 | 1187 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |