Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026815.2 | NP_081091.1 | 6 - 7 | 673 | 65 | |
GenBank mRNA | AF262335.1 | - | 5 - 6 | 645 | 65 | |
AK004014.1 | - | 6 - 7 | 674 | 65 | ||
BC132047.1 | - | 5 - 6 | 665 | 40130324 | 65 | |
BC138390.1 | - | 5 - 6 | 665 | 8861410 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |