Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008408.4 | NP_032434.3 | 10 - 11 | 1187 | 92 | |
GenBank mRNA | AK030363.1 | - | 10 - 11 | 1251 | 92 | |
AK077877.1 | - | 10 - 11 | 1211 | 92 | ||
AK081547.1 | - | 10 - 11 | 1244 | 92 | ||
AK129027.1 | - | 10 - 11 | 1225 | 92 | ||
AK150640.1 | - | 10 - 11 | 1232 | 92 | ||
AK151541.1 | - | 10 - 11 | 1243 | 92 | ||
AK151791.1 | - | 10 - 11 | 1243 | 92 | ||
AK152242.1 | - | 10 - 11 | 1232 | 92 | ||
AK153840.1 | - | 10 - 11 | 1188 | 92 | ||
AK153886.1 | - | 10 - 11 | 1288 | 92 | ||
AK160037.1 | - | 10 - 11 | 1226 | 92 | ||
AK168505.1 | - | 10 - 11 | 1233 | 92 | ||
AK168937.1 | - | 10 - 11 | 1232 | 92 | ||
AK169669.1 | - | 10 - 11 | 1211 | 92 | ||
AX799090.1 | - | 10 - 11 | 1230 | 92 | ||
BC028897.1 | - | 9 - 10 | 1138 | 92 | ||
BC037612.1 | - | 10 - 11 | 1226 | 5353797 | 92 | |
BC085313.1 | - | 10 - 11 | 1216 | 30443880 | 92 | |
L34260.1 | - | 10 - 11 | 1230 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |