Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022813.3 | NP_073724.1 | 4 - 5 | 451 | 59 | |
GenBank mRNA | AF295402.1 | - | 4 - 5 | 455 | 59 | |
AK003080.1 | - | 4 - 5 | 453 | 59 | ||
AK171766.1 | - | 4 - 5 | 442 | 59 | ||
BC014751.1 | - | 4 - 5 | 428 | 4235556 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |