Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104616.1 | NP_001098086.1 | 9 - 10 | 1101 | 131 | |
NM_001104617.1 | NP_001098087.1 | 9 - 10 | 1507 | 131 | ||
NM_009041.3 | NP_033067.2 | 9 - 10 | 1507 | 131 | ||
GenBank mRNA | AK016711.1 | - | 9 - 10 | 1177 | 131 | |
AK146423.1 | - | 9 - 10 | 1165 | 131 | ||
AK156370.1 | - | 9 - 10 | 1193 | 131 | ||
AK162204.1 | - | 9 - 10 | 1079 | 131 | ||
AK168449.1 | - | 9 - 10 | 1178 | 131 | ||
BC034438.1 | - | 9 - 10 | 1333 | 131 | ||
BC053417.1 | - | 9 - 10 | 1181 | 6707873 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |