Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028040.2 | NP_082316.1 | 1 - 2 | 201 | 66 | |
GenBank mRNA | AK010322.1 | - | 1 - 2 | 218 | 66 | |
AK156574.1 | - | 1 - 2 | 218 | 66 | ||
BC061256.1 | - | 1 - 2 | 202 | 30251586 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |