Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK031141.1 | - | 7 - 8 | 750 | 92 | |
AK158516.1 | - | 5 - 6 | 565 | 92 | ||
BC017156.1 | - | 7 - 8 | 871 | 4911201 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |