Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144922.2 | NP_659171.1 | 8 - 9 | 1357 | 74 | |
GenBank mRNA | AK087545.1 | - | 8 - 9 | 1327 | 74 | |
AK143666.1 | - | 8 - 9 | 1400 | 74 | ||
AK155604.1 | - | 8 - 9 | 1368 | 74 | ||
BC021506.1 | - | 8 - 9 | 1381 | 5347099 | 74 | |
BC027844.1 | - | 5 - 6 | 650 | 74 | ||
BC078642.1 | - | 8 - 9 | 1281 | 6329630 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |