Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138951.1 | NP_620401.1 | 1 - 2 | 241 | 65 | |
GenBank mRNA | AK149384.1 | - | 1 - 2 | 272 | 65 | |
AK165811.1 | - | 1 - 2 | 139 | 65 | ||
BC021608.1 | - | 1 - 2 | 153 | 5099426 | 65 | |
BC021613.1 | - | 1 - 2 | 247 | 5102393 | 65 | |
BC024432.1 | - | 1 - 2 | 241 | 4972715 | 65 | |
BC026825.1 | - | 1 - 2 | 188 | 4240199 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |