Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013766.2 | NP_001013788.2 | 4 - 5 | 500 | 84 | |
NM_001115154.1 | NP_001108626.1 | 4 - 5 | 500 | 84 | ||
GenBank mRNA | AK082201.1 | - | 4 - 5 | 501 | 84 | |
BC150876.1 | - | 4 - 5 | 501 | 9087787 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |