Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4x1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003947.1 | NP_001003947.1 | 1 - 2 | 181 | 128 | |
GenBank mRNA | AJ786769.1 | - | 1 - 2 | 181 | 128 | |
AK038526.1 | - | 157 | 128 | |||
BC113125.1 | - | 1 - 2 | 180 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |