Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146013.1 | NP_666125.1 | 7 - 8 | 645 | 92 | |
GenBank mRNA | AK028954.1 | - | 7 - 8 | 606 | 92 | |
BC026948.1 | - | 7 - 8 | 645 | 5136664 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |