Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thnsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033929.2 | NP_001029101.1 | 6 - 7 | 1053 | 63 | |
NM_178413.5 | NP_848500.3 | 7 - 8 | 1121 | 63 | ||
GenBank mRNA | AK165463.1 | - | 6 - 7 | 1019 | 63 | |
BC025604.1 | - | 7 - 8 | 1084 | 5250319 | 63 | |
BC051244.1 | - | 6 - 7 | 1015 | 4975661 | 63 | |
BQ042728.1 | - | 3 - 4 | 402 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |