Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024288.2 | NP_077250.2 | 2 - 3 | 779 | 68 | |
GenBank mRNA | AK164384.1 | - | 2 - 3 | 780 | 68 | |
BC050876.1 | - | 2 - 3 | 630 | 6512227 | 68 | |
CF748545.1 | - | 2 - 3 | 453 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |