Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021449.3 | NP_067424.2 | 9 - 10 | 1054 | 130 | |
NM_175357.3 | NP_780566.1 | 9 - 10 | 1057 | 130 | ||
GenBank mRNA | AF229032.1 | - | 9 - 10 | 1009 | 130 | |
AK032836.1 | - | 9 - 10 | 1031 | 130 | ||
AK050557.1 | - | 9 - 10 | 1040 | 130 | ||
AK075959.1 | - | 9 - 10 | 1030 | 130 | ||
AK076144.1 | - | 9 - 10 | 1037 | 130 | ||
AK160219.1 | - | 9 - 10 | 1033 | 130 | ||
BC046967.1 | - | 8 - 9 | 972 | 130 | ||
BC069905.1 | - | 9 - 10 | 1014 | 130 | ||
BC086488.1 | - | 9 - 10 | 1032 | 30443656 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |