Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242358.1 | NP_001229287.1 | 4 - 5 | 627 | 120 | |
NM_001242360.1 | NP_001229289.1 | 4 - 5 | 562 | 120 | ||
NM_027296.3 | NP_081572.1 | 4 - 5 | 562 | 120 | ||
GenBank mRNA | AB063106.1 | - | 4 - 5 | 534 | 120 | |
AK010662.1 | - | 4 - 5 | 533 | 120 | ||
AK011764.1 | - | 4 - 5 | 578 | 120 | ||
AK135791.1 | - | 4 - 5 | 582 | 120 | ||
AK139764.1 | - | 4 - 5 | 543 | 121 | ||
AK152739.1 | - | 4 - 5 | 598 | 120 | ||
AK166804.1 | - | 4 - 5 | 572 | 120 | ||
BC031764.1 | - | 4 - 5 | 528 | 4486420 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |