Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173775.3 | NP_776136.2 | 9 - 10 | 1077 | 66 | |
GenBank mRNA | AK048790.1 | - | 3 - 4 | 1694 | 66 | |
BC047211.1 | - | 7 - 8 | 759 | 6392183 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |