Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145937.3 | NP_666049.2 | 7 - 8 | 978 | 61 | |
GenBank mRNA | AK151874.1 | - | 7 - 8 | 979 | 61 | |
AK160917.1 | - | 7 - 8 | 960 | 61 | ||
AK161203.1 | - | 7 - 8 | 971 | 61 | ||
BC026981.1 | - | 7 - 8 | 951 | 5365580 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |