Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025358.3 | NP_079634.2 | 2 - 3 | 255 | 99 | |
GenBank mRNA | AK003137.1 | - | 2 - 3 | 237 | 99 | |
AK003214.1 | - | 2 - 3 | 228 | 99 | ||
AK012366.1 | - | 2 - 3 | 235 | 99 | ||
BC005760.1 | - | 1 - 2 | 136 | 99 | ||
BC058378.1 | - | 2 - 3 | 223 | 6820787 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |