Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009650.2 | NP_033780.2 | 2 - 3 | 248 | 89 | |
GenBank mRNA | AF093406.1 | - | 2 - 3 | 174 | 90 | |
BC100458.1 | - | 2 - 3 | 121 | 30917596 | 89 | |
CN837067.1 | - | 2 - 3 | 251 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |