Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Renbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023132.3 | NP_075621.3 | 3 - 4 | 321 | 90 | |
GenBank mRNA | AK014347.1 | - | 3 - 4 | 241 | 90 | |
AK146652.1 | - | 3 - 4 | 286 | 90 | ||
AY183138.1 | - | 3 - 4 | 289 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |