Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194339.1 | NP_919320.1 | 6 - 7 | 938 | 69 | |
GenBank mRNA | AK028266.1 | - | 6 - 7 | 937 | 69 | |
AK129082.1 | - | 6 - 7 | 898 | 69 | ||
AK133013.1 | - | 6 - 7 | 888 | 69 | ||
AK134315.1 | - | 6 - 7 | 946 | 69 | ||
AK135031.1 | - | 5 - 6 | 1435 | 69 | ||
AK162178.1 | - | 6 - 7 | 996 | 69 | ||
BC057054.1 | - | 6 - 7 | 938 | 6827225 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |