Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136227.1 | NP_001129699.1 | 3 - 4 | 516 | 122 | |
NM_009106.2 | NP_033132.2 | 4 - 5 | 923 | 122 | ||
NM_133641.2 | NP_598396.2 | 3 - 4 | 1427 | 122 | ||
GenBank mRNA | AK076155.1 | - | 3 - 4 | 437 | 122 | |
BC004824.1 | - | 3 - 4 | 757 | 122 | ||
BC013820.1 | - | 3 - 4 | 1401 | 3981927 | 122 | |
U54638.1 | - | 4 - 5 | 926 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |