Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008638.2 | NP_032664.1 | 4 - 5 | 642 | 66 | |
GenBank mRNA | AK076019.1 | - | 4 - 5 | 595 | 66 | |
AK152249.1 | - | 4 - 5 | 634 | 66 | ||
AK159680.1 | - | 4 - 5 | 615 | 66 | ||
AK165840.1 | - | 4 - 5 | 634 | 66 | ||
AK166262.1 | - | 4 - 5 | 626 | 66 | ||
AK196703.1 | - | 1 - 2 | 86 | 66 | ||
AK202776.1 | - | 1 - 2 | 85 | 66 | ||
BC019511.1 | - | 4 - 5 | 629 | 4164253 | 66 | |
J04627.1 | - | 4 - 5 | 585 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |