Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077705.2 | NP_001071173.1 | 13 - 14 | 1777 | 87 | |
NM_013545.3 | NP_038573.2 | 13 - 14 | 1775 | 87 | ||
GenBank mRNA | AK132509.1 | - | 13 - 14 | 1694 | 87 | |
AK150509.1 | - | 13 - 14 | 1682 | 87 | ||
AK151077.1 | - | 13 - 14 | 1696 | 87 | ||
AK151623.1 | - | 13 - 14 | 1699 | 87 | ||
AK159686.1 | - | 13 - 14 | 1734 | 87 | ||
AK159833.1 | - | 13 - 14 | 1727 | 87 | ||
AK167923.1 | - | 13 - 14 | 1729 | 87 | ||
AK169804.1 | - | 13 - 14 | 1777 | 87 | ||
BC012660.1 | - | 13 - 14 | 1720 | 4010462 | 87 | |
M68902.1 | - | 13 - 14 | 1727 | 87 | ||
M90389.1 | - | 13 - 14 | 1677 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |