Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028273.1 | - | 4 - 5 | 1094 | 69 | |
AK031129.1 | - | 4 - 5 | 1074 | 69 | ||
AK163571.1 | - | 4 - 5 | 1093 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |